Learn More
Aviva Systems Biology Corp PANK2 Antibody - C-terminal region (ARP63793_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63793_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PANK2. It was validated on Western Blot. . Alternate names: C20orf48, FLJ17232, HARP, HSS, MGC15053, NBIA1, PKANApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-088