Learn More
Medchemexpress LLC HY-10224 500mg , Panobinostat LBH589;NVP-LBH589 CAS:404950-80-7 Purity:98%
SDP
Catalog No. 501492327
Encompass_Preferred
Medchemexpress, HY-10224 500mg Panobinostat LBH589;NVP-LBH589 CAS:404950-80-7 Formula:C21H23N3O2 Purity:98% Panobinostat is an oral histone deacetylase ( HDAC ) inhibitor with a multi-targeted profile. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2327 Supplier Medchemexpress LLC Supplier No. HY10224500MG
Medchemexpress, HY-10224 500mg Panobinostat LBH589;NVP-LBH589 CAS:404950-80-7 Formula:C21H23N3O2 Purity:98% Panobinostat is an oral histone deacetylase ( HDAC ) inhibitor with a multi-targeted profile. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.