Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Panobinostat | 404950-80-7 | MFCD09833242 | 99.4% | 349.4 g/mol | C21H23N3O2 | 10 MG
SDP

Catalog No. 5000245918
Encompass_Preferred

Panobinostat is a non-selective histone deacetylase (HDAC) inhibitor provided as an analytical reference standard for research and analytical use. It is intended for assay calibration, method development, and use as a reference material in studies of HDAC inhibition.

  • Analytical reference standard for panobinostat (LBH589).
  • Purity 99.4%.
  • CAS 404950-80-7.
  • Chemical formula C21H23N3O2; molecular weight 349.4 g/mol.
  • Supplied as a 10 mg solid.
  • Intended for research use only; follow the safety data sheet for handling and storage.

Catalog No. 50-002-45918 Supplier Medchemexpress LLC Supplier No. HY10224R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Panobinostat is a non-selective histone deacetylase (HDAC) inhibitor provided as an analytical reference standard for research and analytical use. It is intended for assay calibration, method development, and use as a reference material in studies of HDAC inhibition.

  • Analytical reference standard for panobinostat (LBH589).
  • Purity 99.4%.
  • CAS 404950-80-7.
  • Chemical formula C21H23N3O2; molecular weight 349.4 g/mol.
  • Supplied as a 10 mg solid.
  • Intended for research use only; follow the safety data sheet for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.