Learn More
Medchemexpress LLC Panobinostat | 404950-80-7 | MFCD09833242 | 99.4% | 349.43 | C21H23N3O2 | 200 MG
SDP
Catalog No. 5000245554
Encompass_Preferred
Panobinostat is a potent, orally active, non-selective histone deacetylase (HDAC) inhibitor supplied as a research-grade powder for laboratory use. It exhibits antineoplastic activity and is commonly used in epigenetic and cancer research, and in studies of apoptosis and autophagy. Not for human or veterinary use.
  • Potent non-selective HDAC inhibitor with antineoplastic activity.
  • High purity (99.4% by LCMS).
  • Powder form, soluble in DMSO ≥ 100 mg/mL.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (1 year).
  • Used in studies of apoptosis, autophagy, and cancer cell proliferation.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-45554 Supplier Medchemexpress LLC Supplier No. HY10224200MG
Panobinostat is a potent, orally active, non-selective histone deacetylase (HDAC) inhibitor supplied as a research-grade powder for laboratory use. It exhibits antineoplastic activity and is commonly used in epigenetic and cancer research, and in studies of apoptosis and autophagy. Not for human or veterinary use.
  • Potent non-selective HDAC inhibitor with antineoplastic activity.
  • High purity (99.4% by LCMS).
  • Powder form, soluble in DMSO ≥ 100 mg/mL.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (1 year).
  • Used in studies of apoptosis, autophagy, and cancer cell proliferation.
  • Intended for research use only; not for human or veterinary use.