Learn More
Medchemexpress LLC HY-10224 50mg , Panobinostat CAS:404950-80-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1022450MG
Encompass_Preferred
HY-10224 50mg Panobinostat CAS: 404950-80-7 Panobinostat (LBH589; NVP-LBH589) is a potent and orally active non-selective HDAC inhibitor, and has antineoplastic activities. Panobinostat induces HIV-1 virus production even at low concentration range 8-31 nM, stimulates HIV-1 expression in latently infected cells[4]. Panobinostat induces cell apoptosis and autophagy. Panobinostat can be used for the study of refractory or relapsed multiple myeloma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8998