Learn More
Hillrom PanOptic Ophthalmoscope Lamp
Catalog No. 22355152
Change view
Click to view available options
Product Type:
Halogen Lamp
Catalog No. Product Type
22-355-152 Halogen Lamp
1 options
This item is not returnable. View return policy
Catalog No. 22-355-152 Supplier Hillrom Supplier No. WEL 03800U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Halogen HPX Lamp for PanOptic Ophthalmoscope provides bright white light. Consistent and reliable usage.
  • Halogen Lamp
  • For use with PanOptic Ophthalmoscope 11810, 11820, and 11800
  • Warranty: 30 days
  • 3.5 V
  • 10 Watts
Specifications
Product Type Halogen Lamp