Learn More
Hach Company Pans, Aluminum, 70 mm, 100/PK
Supplier:  Hach Company 2948000
Encompass
Aluminum pans used in Solid-Phase Extraction analysis, EPA method 1664A. 70mm. 100/pk.
Catalog No. 50-199-6941