Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Panserin 293A, Serum-free medium for HEK-Cells in adherent culture, w: L-Glutamine (500 ml)
SDP
Catalog No. 502388490
Encompass_Preferred
PAN-Biotech Panserin 293A, Serum-free medium for HEK-Cells in adherent culture, w: L-Glutamine. QUANTITY: 500 ml. DESCRIPTION: Panserin 293A is a ready-to-use medium for the serum-free cultivation of HEK293 cells (Human Embryonic Kidney) in adherent culture. Serum-containing medium can be replaced directly. Panserin 293A promotes cell attachment and high cell growth rates, and has been used for the expression of recombinant proteins and the proliferation of AVVs (Adeno-associated virus vectors) and LVVs (lentiviral vectors). The efficient serum-free cultivation can be supported by higher seeding densities. COMPOSITION: Based on DMEM, Panserin 293A contains additional trace elements, proteins, cholesterol, lipids, vitamins and hormones. The total content of proteins and animal-derived components is lower than 0.2% w/v.
Catalog No. 50-238-8490 Supplier ILEX LIFE SCIENCES LLC Supplier No. P04710608
PAN-Biotech Panserin 293A, Serum-free medium for HEK-Cells in adherent culture, w: L-Glutamine. QUANTITY: 500 ml. DESCRIPTION: Panserin 293A is a ready-to-use medium for the serum-free cultivation of HEK293 cells (Human Embryonic Kidney) in adherent culture. Serum-containing medium can be replaced directly. Panserin 293A promotes cell attachment and high cell growth rates, and has been used for the expression of recombinant proteins and the proliferation of AVVs (Adeno-associated virus vectors) and LVVs (lentiviral vectors). The efficient serum-free cultivation can be supported by higher seeding densities. COMPOSITION: Based on DMEM, Panserin 293A contains additional trace elements, proteins, cholesterol, lipids, vitamins and hormones. The total content of proteins and animal-derived components is lower than 0.2% w/v.