Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Panserin 293S, Serum-free medium for HEK-Cells in suspension culture, w: L-Glutamine (2 x 500 ml)
SDP
Catalog No. 502388473
Encompass_Preferred
PAN-Biotech Panserin 293S: Serum-free medium for HEK293 cells in suspension culture, w: L-Glutamine. QUANTITY: 2 x 500 ml. MANUFACTURED BY: Pan-Biotech (Germany). DESCRIPTION: PAN-Biotech Panserin 293S is a complete, ready to use medium for the serum-free cultivation of HEK293 cells (Human Embryonic Kidney) in suspension (non-adherent) culture. COMPOSITION: Based on DMEM/F12 medium with additional oligopeptides (hydrolysates < 0.02% w/v), hormones and trace elements to support expression and cell growth. This product contains only traces of protein (< 0.002% w/v) and animal derived components (< 0.0003% w/v). APPLICATION: Panserin 293S is a particularly enriched medium optimized for the growth of HEK293 cells in suspension culture. Due to its defined composition, the purification of final products (recombinant proteins, viruses) from the cell culture is more convenient and economic. This product can also be applied to adapt adherent HEK293 cells into suspension.
Catalog No. 50-238-8473 Supplier ILEX LIFE SCIENCES LLC Supplier No. P04710609X2
PAN-Biotech Panserin 293S: Serum-free medium for HEK293 cells in suspension culture, w: L-Glutamine. QUANTITY: 2 x 500 ml. MANUFACTURED BY: Pan-Biotech (Germany). DESCRIPTION: PAN-Biotech Panserin 293S is a complete, ready to use medium for the serum-free cultivation of HEK293 cells (Human Embryonic Kidney) in suspension (non-adherent) culture. COMPOSITION: Based on DMEM/F12 medium with additional oligopeptides (hydrolysates < 0.02% w/v), hormones and trace elements to support expression and cell growth. This product contains only traces of protein (< 0.002% w/v) and animal derived components (< 0.0003% w/v). APPLICATION: Panserin 293S is a particularly enriched medium optimized for the growth of HEK293 cells in suspension culture. Due to its defined composition, the purification of final products (recombinant proteins, viruses) from the cell culture is more convenient and economic. This product can also be applied to adapt adherent HEK293 cells into suspension.