Learn More
ILEX LIFE SCIENCES LLC PANSERIN 701 SFM 500 ML
SDP
Catalog No. 502733775
Encompass_Preferred
PAN-Biotech Panserin 701, Serum-free medium for Lymphocytes, w: L-Glutamine, w: PHA-L (500 ml). MANUFACTURER: PAN-Biotech GmbH (Germany). INTRO: PAN-Biotech Panserin 701 is a fully defined, complete ready-to-use serum-free medium for the cultivation of lymphocytes from whole blood. CELL TYPE: Lymphocytes (T cells, B cells, natural killer [NK] cells). CULTURE TYPE: Suspension. GLUTAMINE: With L-glutamine. STERILE: Yes. STORAGE TEMP: -20°C. COMPOSITION: Based on Iscove's MEM, the medium is enriched with additional trace elements, albumin, cholesterol, lipids and vitamins. It contains the mitogen phytohemagglutinin (PHA) for the growth stimulation of lymphocytes. This product contains traces of animal-derived components (< 0.2% w/v) and proteins (< 0.2% w/v). APPLICATION: Panserin 701 has been developed for the serum-free cultivation of lymphocytes from whole blood. The plant lectin (PHA) in Panserin 701 stimulates cell division.
Catalog No. 50-273-3775 Supplier ILEX LIFE SCIENCES LLC Supplier No. P04710701
Add to Cart
Add to Cart
PAN-Biotech Panserin 701, Serum-free medium for Lymphocytes, w: L-Glutamine, w: PHA-L (500 ml). MANUFACTURER: PAN-Biotech GmbH (Germany). INTRO: PAN-Biotech Panserin 701 is a fully defined, complete ready-to-use serum-free medium for the cultivation of lymphocytes from whole blood. CELL TYPE: Lymphocytes (T cells, B cells, natural killer [NK] cells). CULTURE TYPE: Suspension. GLUTAMINE: With L-glutamine. STERILE: Yes. STORAGE TEMP: -20°C. COMPOSITION: Based on Iscove's MEM, the medium is enriched with additional trace elements, albumin, cholesterol, lipids and vitamins. It contains the mitogen phytohemagglutinin (PHA) for the growth stimulation of lymphocytes. This product contains traces of animal-derived components (< 0.2% w/v) and proteins (< 0.2% w/v). APPLICATION: Panserin 701 has been developed for the serum-free cultivation of lymphocytes from whole blood. The plant lectin (PHA) in Panserin 701 stimulates cell division.