Learn More
Med Vet International Pantarsal Arthrodesis Plate, Feline, Left, 78mm, J0433CL, Each
Catalog No. 5000582594
Encompass_Preferred
Our pantarsal plate is 135° with the middle screw placed in the center of the talus or the center of motion of the joint. This is not a one size fits all, but the only alternative is a custom plate. For cats. Overall length 78mm. 120° angle, better to match feline anatomy especially useful for severe shearing injuries. 2.7mm/2.0mm screws.
Catalog No. 50-005-82594 Supplier Med Vet International Supplier No. JOR433CL
Our pantarsal plate is 135° with the middle screw placed in the center of the talus or the center of motion of the joint. This is not a one size fits all, but the only alternative is a custom plate. For cats. Overall length 78mm. 120° angle, better to match feline anatomy especially useful for severe shearing injuries. 2.7mm/2.0mm screws.