Learn More
Med Vet International Pantarsal Arthrodesis Plate, Left, 2.7 / 3.5mm, J0433A, Each
Catalog No. 5000582681
Encompass_Preferred
The surgical immobilization of the hock joint can be difficult. A medially applied plate appears to best neutralize the effect of the long lever arm created by arthrodesis. Data from the manufacture of 50 custom plates has enabled JorVet to create a pantarsal plate which fits most medium and large dogs. The proximal portion is standard 3.5mm DCP, broadening over the vulnerable calcaneoquartal region, and finally tapering out over the metatarsals as a 2.7mm DCP.
Catalog No. 50-005-82681 Supplier Med Vet International Supplier No. JOR433A
The surgical immobilization of the hock joint can be difficult. A medially applied plate appears to best neutralize the effect of the long lever arm created by arthrodesis. Data from the manufacture of 50 custom plates has enabled JorVet to create a pantarsal plate which fits most medium and large dogs. The proximal portion is standard 3.5mm DCP, broadening over the vulnerable calcaneoquartal region, and finally tapering out over the metatarsals as a 2.7mm DCP.