Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Panthenol, USP, 99-102%, Spectrum™ Chemical
SDP

Catalog No. 18604971 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-971 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-971 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P191012KGBL

missing translation for 'validMainframeMsg'

P1910, 16485-10-2, C9H19NO4

Spectrum™ Chemical Panthenol, USP is used as a topical moisturizer and lubricant to treat skin diorders. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 16485-10-2
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00002944
InChI Key SNPLKNRPJHDVJA-UHFFFAOYNA-N
IUPAC Name 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)C(O)C(=O)NCCCO

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 64.5-68.5°C
Packaging Poly Pail
Quantity 12 kg
Specific Rotation -0.05° to +0.05°
Percent Purity 99 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.