Learn More
Motic™ Panthera E2 Compound LED Microscopes
GREENER_CHOICE
Catalog No. 01258020 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Binocular
Trinocular
Magnification Power:
4X, 10X, 40XS, 100XS (oil)
Eyepiece:
WF 10X/20 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style Magnification Power Eyepiece
01-258-020 Binocular 4X, 10X, 40XS, 100XS (oil) WF 10X/20 mm
01-258-041 Trinocular 4X, 10X, 40XS, 100XS (oil) WF 10X/20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-258-020 Supplier Motic™ Supplier No. 1100104601116
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for the future, the Panthera E2 is ready for ambitious work in educational environments and small life science labs. Features new CCIS™ Plan Ultra Contrast Objectives with stunning image quality in Brightfield, Phase Contrast, Darkfield, and Polarization.
  • Swiveling, 25° inclined Siedentopf head
  • LED light indicator
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built-in, low position coaxial rackless stage with right hand control
  • Focusable Abbe condenser with 0.9 numerical aperture
  • Iris diaphragm and slot
  • Diopter Adjustment: ±5
Specifications
Eyepiece WF 10X/20 mm
Head Style Binocular
Lighting Type LED
Magnification Power 4X, 10X, 40XS, 100XS (oil)
Model Panthera E2 Binocular
Objective Types Infinity Corrected Plan Achromatic
Product Line Panthera
Product Type Compound Microscope
Nosepiece Reversed Quadruple
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov