Learn More
ATCC Lactobacillus acidophilus
Catalog No. 502379982
Encompass
Lactobacillus acidophilus strain [43] is a quality control strain. This bacterium is propagated in a 5% CO2 atmosphere on lactobacilli MRS medium.
Catalog No. 50-237-9982 Supplier ATCC Supplier No. 314
Lactobacillus acidophilus strain [43] is a quality control strain. This bacterium is propagated in a 5% CO2 atmosphere on lactobacilli MRS medium.