Learn More
Medchemexpress LLC Pantoprazole | 102625-70-7 | 99.9% | 383.37 | 100 MG
SDP
Catalog No. 5000386310
Encompass_Preferred
Pantoprazole is an orally active and potent proton pump inhibitor (PPI). It is a substituted benzimidazole and a potent H+/K+-ATPase inhibitor. It improves pH stability and has anti-secretory and anti-ulcer activities. This product is intended for research and analytical applications.
  • Improves pH stability
  • Has anti-secretory activities
  • Has anti-ulcer activities
  • Potent H+/K+-ATPase inhibitor
Catalog No. 50-003-86310 Supplier Medchemexpress LLC Supplier No. HY17507100MG
Pantoprazole is an orally active and potent proton pump inhibitor (PPI). It is a substituted benzimidazole and a potent H+/K+-ATPase inhibitor. It improves pH stability and has anti-secretory and anti-ulcer activities. This product is intended for research and analytical applications.
  • Improves pH stability
  • Has anti-secretory activities
  • Has anti-ulcer activities
  • Potent H+/K+-ATPase inhibitor