Learn More
Medchemexpress LLC Pantoprazole | 102625-70-7 | 99.95% | 383.37 | 1 ML
SDP
Catalog No. 5000386214
Encompass_Preferred
Pantoprazole (BY10232) is an orally active and potent proton pump inhibitor (PPI). It is a substituted benzimidazole and a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. Pantoprazole improves pH stability and possesses anti-secretory and anti-ulcer activities. It also significantly increased tumor growth delay when combined with Doxorubicin.
  • Orally active and potent proton pump inhibitor (PPI)
  • Substituted benzimidazole
  • Potent H+/K+-ATPase inhibitor with an an IC50 of 6.8 μM
  • Improves pH stability
  • Anti-secretory and anti-ulcer activities
  • Increased tumor growth delay in combination with Doxorubicin
Catalog No. 50-003-86214 Supplier Medchemexpress LLC Supplier No. HY1750710MM/1ML
Pantoprazole (BY10232) is an orally active and potent proton pump inhibitor (PPI). It is a substituted benzimidazole and a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. Pantoprazole improves pH stability and possesses anti-secretory and anti-ulcer activities. It also significantly increased tumor growth delay when combined with Doxorubicin.
  • Orally active and potent proton pump inhibitor (PPI)
  • Substituted benzimidazole
  • Potent H+/K+-ATPase inhibitor with an an IC50 of 6.8 μM
  • Improves pH stability
  • Anti-secretory and anti-ulcer activities
  • Increased tumor growth delay in combination with Doxorubicin