Learn More
Selleck Chemical LLC Pantoprazole S2105-500mg
SDP
Catalog No. 503109695
Encompass_Preferred
Pantoprazole (SKF96022 BY-1023) is a proton pump inhibitor drug that inhibits gastric acid secretion It works on gastric parietal cells to irreversibly inhibit (H /K )-ATPase function and suppress the production of gastric acid
Catalog No. 50-310-9695 Supplier Selleck Chemical LLC Supplier No. S2105500MG
Pantoprazole (SKF96022 BY-1023) is a proton pump inhibitor drug that inhibits gastric acid secretion It works on gastric parietal cells to irreversibly inhibit (H /K )-ATPase function and suppress the production of gastric acid