Learn More
Medchemexpress LLC Pantoprazole-d8 sodium | >98.0% | 395.42 g/mol | C16H10D8F2N3O4S | 1mg
SDP
Catalog No. 5000270270
Encompass_Preferred
Pantoprazole-d8 (BY1023-d8) sodium is a deuterium labeled Pantoprazole (HY-17507) Pantoprazole (BY10232) is an orally active and potent proton pump inhibitor (PPI)[1] Pantoprazole a substituted benzimidazole is a potent H /K -ATPase inhibitor with an IC50 of 6 8 M Pantoprazole improves pH stability and has anti-secretory anti-ulcer activities Pantoprazole significantly increased tumor growth delay combined with Doxorubicin (HY-15142)[3][4]
Catalog No. 50-002-70270 Supplier Medchemexpress LLC Supplier No. HYW7397931MG
Pantoprazole-d8 (BY1023-d8) sodium is a deuterium labeled Pantoprazole (HY-17507) Pantoprazole (BY10232) is an orally active and potent proton pump inhibitor (PPI)[1] Pantoprazole a substituted benzimidazole is a potent H /K -ATPase inhibitor with an IC50 of 6 8 M Pantoprazole improves pH stability and has anti-secretory anti-ulcer activities Pantoprazole significantly increased tumor growth delay combined with Doxorubicin (HY-15142)[3][4]