Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pantoprazole impurity 42 | 107512-34-5 | 157.60 | C7H8ClNO | 25 G
SDP

Catalog No. 5000209655
Encompass_Preferred

Pantoprazole impurity 42 is a defined impurity and analytical reference standard related to the proton-pump inhibitor pantoprazole. It is intended for research and analytical use, including method development and impurity profiling.

  • Defined impurity for analytical characterization.
  • Suitable as a reference standard or drug intermediate.
  • CAS number 107512-34-5; molecular formula C7H8ClNO.
  • Molecular weight 157.60 g/mol.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-09655 Supplier Medchemexpress LLC Supplier No. 25GHYW059074
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pantoprazole impurity 42 is a defined impurity and analytical reference standard related to the proton-pump inhibitor pantoprazole. It is intended for research and analytical use, including method development and impurity profiling.

  • Defined impurity for analytical characterization.
  • Suitable as a reference standard or drug intermediate.
  • CAS number 107512-34-5; molecular formula C7H8ClNO.
  • Molecular weight 157.60 g/mol.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.