Learn More
Medchemexpress LLC Pantoprazole sodium | 138786-67-1 | 99.8% | 405.35 | 500 MG
SDP
Catalog No. 5000386384
Encompass_Preferred
Pantoprazole sodium is an orally active and potent proton pump inhibitor (PPI) that improves pH stability and exhibits anti-secretory and anti-ulcer activities. It acts as a potent H+/K+-ATPase inhibitor, also shown to significantly increase tumor growth delay when combined with Doxorubicin.
  • Orally active and potent proton pump inhibitor
  • Potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM
  • Improves pH stability
  • Exhibits anti-secretory and anti-ulcer activities
  • Increases tumor growth delay when combined with Doxorubicin
  • Blocks exosome release
  • Inhibits V-H+-ATPase activity
Catalog No. 50-003-86384 Supplier Medchemexpress LLC Supplier No. HY17507A500MG
Pantoprazole sodium is an orally active and potent proton pump inhibitor (PPI) that improves pH stability and exhibits anti-secretory and anti-ulcer activities. It acts as a potent H+/K+-ATPase inhibitor, also shown to significantly increase tumor growth delay when combined with Doxorubicin.
  • Orally active and potent proton pump inhibitor
  • Potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM
  • Improves pH stability
  • Exhibits anti-secretory and anti-ulcer activities
  • Increases tumor growth delay when combined with Doxorubicin
  • Blocks exosome release
  • Inhibits V-H+-ATPase activity