Learn More
Medchemexpress LLC Pantoprazole sodium | 138786-67-1 | MFCD01658543 | 405.4 g/mol | C16H14F2N3NaO4S | 10 MG
SDP
Catalog No. 5000299137
Encompass_Preferred
Pantoprazole sodium is the sodium salt form of pantoprazole, a substituted benzimidazole proton pump inhibitor used in research to modulate gastric acid secretion and related biological pathways. The sodium salt form offers improved solubility and handling for laboratory preparation and analytical work.
  • Sodium salt form of a proton pump inhibitor suitable for research use.
  • Useful for in vitro and in vivo studies of gastric acid secretion and related pathways.
  • Improved solubility and stability compared with the free base for solution preparation.
  • Suitable as an analytical standard and for assay development.
  • Supplied in laboratory-scale quantities for research applications.
Catalog No. 50-002-99137 Supplier Medchemexpress LLC Supplier No. HY17507AR10MG
Pantoprazole sodium is the sodium salt form of pantoprazole, a substituted benzimidazole proton pump inhibitor used in research to modulate gastric acid secretion and related biological pathways. The sodium salt form offers improved solubility and handling for laboratory preparation and analytical work.
  • Sodium salt form of a proton pump inhibitor suitable for research use.
  • Useful for in vitro and in vivo studies of gastric acid secretion and related pathways.
  • Improved solubility and stability compared with the free base for solution preparation.
  • Suitable as an analytical standard and for assay development.
  • Supplied in laboratory-scale quantities for research applications.