Learn More
Medchemexpress LLC Pantoprazole (sodium hydrate) | 164579-32-2 | 99.9% | 432.37 | 100 MG
SDP
Catalog No. 5000386308
Encompass_Preferred
Pantoprazole sodium hydrate is an orally active and potent proton pump inhibitor. It is a substituted benzimidazole and a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. It improves pH stability and possesses anti-secretory and anti-ulcer activities.
  • Orally active and potent proton pump inhibitor
  • Substituted benzimidazole
  • Potent H+/K+-ATPase inhibitor
  • Improves pH stability
  • Has anti-secretory activity
  • Has anti-ulcer activities
  • Can increase tumor growth delay when combined with Doxorubicin
  • Increases endosomal pH in EMT-6 and MCF7 cells
  • Blocks exosome release
  • Inhibits V-H+-ATPase activity
Catalog No. 50-003-86308 Supplier Medchemexpress LLC Supplier No. HY17507B100MG
Pantoprazole sodium hydrate is an orally active and potent proton pump inhibitor. It is a substituted benzimidazole and a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. It improves pH stability and possesses anti-secretory and anti-ulcer activities.
  • Orally active and potent proton pump inhibitor
  • Substituted benzimidazole
  • Potent H+/K+-ATPase inhibitor
  • Improves pH stability
  • Has anti-secretory activity
  • Has anti-ulcer activities
  • Can increase tumor growth delay when combined with Doxorubicin
  • Increases endosomal pH in EMT-6 and MCF7 cells
  • Blocks exosome release
  • Inhibits V-H+-ATPase activity