Learn More
Medchemexpress LLC Pantoprazole sodium hydrate | 164579-32-2 | 99.9% | 432.37 | 250 MG
SDP
Catalog No. 5000385314
Encompass_Preferred
Pantoprazole sodium hydrate is an orally active and potent proton pump inhibitor (PPI). It is a substituted benzimidazole and a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. It improves pH stability and has anti-secretory and anti-ulcer activities. When combined with Doxorubicin, Pantoprazole sodium hydrate significantly increased tumor growth delay.
  • Potent proton pump inhibitor
  • Improves pH stability
  • Possesses anti-secretory activities
  • Exhibits anti-ulcer activities
  • Can increase tumor growth delay when combined with Doxorubicin
Catalog No. 50-003-85314 Supplier Medchemexpress LLC Supplier No. 250MGHY17507AR
Pantoprazole sodium hydrate is an orally active and potent proton pump inhibitor (PPI). It is a substituted benzimidazole and a potent H+/K+-ATPase inhibitor with an IC50 of 6.8 μM. It improves pH stability and has anti-secretory and anti-ulcer activities. When combined with Doxorubicin, Pantoprazole sodium hydrate significantly increased tumor growth delay.
  • Potent proton pump inhibitor
  • Improves pH stability
  • Possesses anti-secretory activities
  • Exhibits anti-ulcer activities
  • Can increase tumor growth delay when combined with Doxorubicin