Learn More
Selleck Chemical LLC Pantoprazole sodium hydrate
SDP
Catalog No. 5000003819
Encompass_Preferred
Pantoprazole (BY10232 SKF96022) sodium hydrate (Protonix) is a potent inhibitor of H /K( )-ATPase with IC50 of 6 8 M
Catalog No. 50-000-03819 Supplier Selleck Chemical LLC Supplier No. S5608100MG
Pantoprazole (BY10232 SKF96022) sodium hydrate (Protonix) is a potent inhibitor of H /K( )-ATPase with IC50 of 6 8 M