Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pantoprazole sulfide | 102625-64-9 | MFCD07368273 | 98.8% | 367.37 g/mol | C16H15F2N3O3S | 500 G
SDP

Catalog No. 5000303903
Encompass_Preferred

Pantoprazole sulfide is the primary metabolite of pantoprazole, provided as a high-purity research reagent for laboratory use. The compound (C16H15F2N3O3S, MW 367.37 g/mol) is supplied as a powder with documented purity and storage conditions. For research use only; not for human or clinical use.

  • High purity: 98.8%.
  • Molecular formula C16H15F2N3O3S; molecular weight 367.37 g/mol.
  • Available in gram-scale packages, including a 500 g option.
  • Powder form with long-term stability when stored frozen.
  • Storage recommendations provided for powder and solutions.
  • Suitable as a metabolite reference for analytical and metabolic studies.

Catalog No. 50-003-03903 Supplier Medchemexpress LLC Supplier No. HYW016033500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pantoprazole sulfide is the primary metabolite of pantoprazole, provided as a high-purity research reagent for laboratory use. The compound (C16H15F2N3O3S, MW 367.37 g/mol) is supplied as a powder with documented purity and storage conditions. For research use only; not for human or clinical use.

  • High purity: 98.8%.
  • Molecular formula C16H15F2N3O3S; molecular weight 367.37 g/mol.
  • Available in gram-scale packages, including a 500 g option.
  • Powder form with long-term stability when stored frozen.
  • Storage recommendations provided for powder and solutions.
  • Suitable as a metabolite reference for analytical and metabolic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.