Learn More
Aviva Systems Biology Corp PANX2 Antibody - N-terminal region (ARP42778_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42778_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PANX2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PANX2. Alternate names: hPANX2, PX2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53351