Learn More
Hach Company Phenylarsine Oxide (PAO) Digital Titrator Cartridge, 0.00451 N
Catalog No. 501995654 Shop All Hach Company Products
Encompass
For chlorine determination by Digital Titrator titration. 0.00451 N PAO. 1 digit = 1µg/L CI2 in 200 mL sample. Digits x 0.00000564 = meq.
Catalog No. 50-199-5654 Supplier Hach Company Supplier No. 2259901
For chlorine determination by Digital Titrator titration. 0.00451 N PAO. 1 digit = 1µg/L CI2 in 200 mL sample. Digits x 0.00000564 = meq.