Learn More
Hach Company Phenylarsine Oxide (PAO) Digital Titrator Cartridge, 0.00564 N
Supplier:  Hach Company 199901
Encompass
For low level chlorine determination by amperometric titration, using the Digital Titrator
Catalog No. 50-199-4653