Learn More
Hach Company Phenylarsine Oxide (PAO) Digital Titrator Cartridge, 0.0451 N
Catalog No. 501995471 Shop All Hach Company Products
Encompass
For chlorine determination by Digital Titrator titraton. 0.00451 N. 1 digit = 1 mg/L CI2 in 200 mL sample, Digits x 0.00000564 = meq.
Catalog No. 50-199-5471 Supplier Hach Company Supplier No. 2142001
For chlorine determination by Digital Titrator titraton. 0.00451 N. 1 digit = 1 mg/L CI2 in 200 mL sample, Digits x 0.00000564 = meq.