Learn More
Hach Company Phenylarsine Oxide (PAO) Titrant, 0.00564N, 500 mL
Supplier:  Hach Company 2700349
Encompass
For Free and Total Chlorine determination by amperometric titration. 0.00564 N PAO. 500 mL bottle.
Catalog No. 50-199-6572