Learn More
Aviva Systems Biology Corp PAOX Antibody - C-terminal region (ARP53829_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53829_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAOX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PAOX. Alternate names: RP11-122K13.11, DKFZp434J245, MGC45464, PAOApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61141