Learn More
Medchemexpress LLC PAP 248-286 (acetate) | 98.9% | 4551.39 | 1 MG
SDP
Catalog No. 5000398902
Encompass_Preferred
PAP 248-286 (Prostatic acid phosphatase (248-286)) acetate is a biologically active peptide. It is a semen-derived enhancer of viral infection (SEVI) factor found in semen. This peptide significantly increases HIV infection by enhancing virion attachment to target cells.
  • Biologically active peptide
  • Semen-derived enhancer of viral infection (SEVI) factor
  • Enhances virion attachment to target cells, increasing HIV infection
  • Potential applications in COVID-19 anti-viral research
Catalog No. 50-003-98902 Supplier Medchemexpress LLC Supplier No. HYP5363A5MG
PAP 248-286 (Prostatic acid phosphatase (248-286)) acetate is a biologically active peptide. It is a semen-derived enhancer of viral infection (SEVI) factor found in semen. This peptide significantly increases HIV infection by enhancing virion attachment to target cells.
  • Biologically active peptide
  • Semen-derived enhancer of viral infection (SEVI) factor
  • Enhances virion attachment to target cells, increasing HIV infection
  • Potential applications in COVID-19 anti-viral research