Learn More
Enzo Life Sciences PAP-AMC, Tyrosinase substrate (0.1mg)
SDP
Supplier:  Enzo Life Sciences BMLEI4050100
Encompass_Preferred
Fluorogenic substrate for tyrosinase. When assaying in phosphate buffer pH 7.1, the optimal excitation/emission wavelengths are 340/440 nm. 325/460 nm also work, but 380/460 nm will not detect this substrate. Alternative name: 1-(4-Methyl-coumarin-7-yl)-3-(4-hydroxyphenyl)urea. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (18 mg/ml) or in methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0783