Learn More
Abcam PAP-AMC, Tyrosinase substrate, Fluorogenic tyrosinase substrate, 100UG
Catalog No. 502705392
Encompass_Preferred
MW 310.3 g/mol, Purity >98%. Fluorogenic tyrosinase substrate. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5392 Supplier Abcam Supplier No. AB145211100UG
MW 310.3 g/mol, Purity >98%. Fluorogenic tyrosinase substrate. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement