Learn More
Drg International Inc Pap Elisa Ruo
Supplier:  Drg International Inc EIA3763R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of plasmin-alpha2-antiplasmin complex (PAP) in plasma (EDTA or citrate plasma). This product is for Research Use Only. The DRG PAP ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. During the first incubation step the PAP present in the sample binds to the monoclonal antibodies (PAP-6) coated on the surface of the microtitration plate.
Catalog No. 50-194-7599