Learn More
Abcam PAP pen, 1UNIT
Catalog No. NC9063266
Encompass_Preferred
Hydrophobic barrier PAP pen (pH 5.8) is a special marking pen that provides a thin film-like green-tinged barrier when a circle is drawn gently around a specimen on a slide. - Cited in over 15 publications - Good for approximately 500 uses - Contains 35% toluene, 15% solvent naphtha, 15% xylene - Insoluble in alcohol and acetone
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC9063266 Supplier Abcam Supplier No. AB26011UNIT
Hydrophobic barrier PAP pen (pH 5.8) is a special marking pen that provides a thin film-like green-tinged barrier when a circle is drawn gently around a specimen on a slide. - Cited in over 15 publications - Good for approximately 500 uses - Contains 35% toluene, 15% solvent naphtha, 15% xylene - Insoluble in alcohol and acetone
The product is subject to the following: Abcam Restricted Use Statement