Learn More
Sigma Aldrich Fine Chemicals Biosciences PAP pen for immunostaining, 5 mm tip
Catalog No. NC0549152
Encompass
Use pen to draw a hydrophobic circle around slide-mounted tissue. Prevents the waste of valuable reagents by keeping liquid pooled in a small droplet. Insoluble in ethanol, acetone and water, but will wash off with xylene.
Catalog No. NC0549152 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3778211EA
Use pen to draw a hydrophobic circle around slide-mounted tissue. Prevents the waste of valuable reagents by keeping liquid pooled in a small droplet. Insoluble in ethanol, acetone and water, but will wash off with xylene.