Learn More
Scytek Laboratories Pap Pen, Draw Film-Like Barrier around Specimen
SDP
Catalog No. NC9401632
Encompass_Preferred
Pen; Pap; Designed to provide a thin film-like barrier when a circle is drawn around the specimen on a slide; barrier creates proper surface tension to hold an antibody solution within the target area on the slide; special formulation
Catalog No. NC9401632 Supplier Scytek Laboratories Supplier No. LP0001
Pen; Pap; Designed to provide a thin film-like barrier when a circle is drawn around the specimen on a slide; barrier creates proper surface tension to hold an antibody solution within the target area on the slide; special formulation