Learn More
Scytek Laboratories PAP Pen Mini, Marking Pen, Insoluble in Alcohol and Acetone
SDP
Catalog No. NC9401633
Encompass_Preferred
This marking pen has been designed to provide a thin film-like barrier when a circle is drawn around the specimen on a slide. This barrier creates the proper surface tension to hold an antibody solution within the target area on the slide. PAP Pen contains a special formulation which is insoluble in alcohol and acetone. It can be removed, if desired, by xylene after the staining procedure is completed.
Catalog No. NC9401633 Supplier Scytek Laboratories Supplier No. LP0002
This marking pen has been designed to provide a thin film-like barrier when a circle is drawn around the specimen on a slide. This barrier creates the proper surface tension to hold an antibody solution within the target area on the slide. PAP Pen contains a special formulation which is insoluble in alcohol and acetone. It can be removed, if desired, by xylene after the staining procedure is completed.