Learn More
Cayman Chemical PAPA NONOate, Purity: ≥98%, CAS: 146672-58-4, 100 mg
SDP
Catalog No. 502892483
Encompass_Preferred
PAPA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 15 minutes and 77 minutes at 37°C and 22-25°C, respectively, pH 7.4, to liberate 2 moles of NO per mole of parent compound.{1190,5437}
Catalog No. 50-289-2483 Supplier Cayman Chemical Supplier No. 82140100 MG
PAPA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 15 minutes and 77 minutes at 37°C and 22-25°C, respectively, pH 7.4, to liberate 2 moles of NO per mole of parent compound.{1190,5437}