Learn More
Enzo Life Sciences PAPA NONOate (25mg). CAS: 146672-58-4
SDP
Catalog No. 502009146
Encompass_Preferred
Nitric oxide (NO) donor. The half-life time of PAPA NONOate is 76.6 min. at 22°C in 0.1M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0. Alternative name: PAPA/NO, NOC-15, (Z)-1-[N-(3-Ammoniopropyl)-N-(n-propyl)amino]diazen-1-ium-1,2-diolate. Purity: ≥97% (TLC). Appearance: White to off-white solid. Solubility: Soluble in water or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9146 Supplier Enzo Life Sciences Supplier No. ALX430016M025
Nitric oxide (NO) donor. The half-life time of PAPA NONOate is 76.6 min. at 22°C in 0.1M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0. Alternative name: PAPA/NO, NOC-15, (Z)-1-[N-(3-Ammoniopropyl)-N-(n-propyl)amino]diazen-1-ium-1,2-diolate. Purity: ≥97% (TLC). Appearance: White to off-white solid. Solubility: Soluble in water or methanol. Long Term Storage: -20°C.