Learn More
Bioworld Peptone S (Soy Peptone), Papaic Digest of Soybean Meal, 500 g
Catalog No. 50254349
Encompass_Preferred
Papaic Digest of Soybean Meal (Peptone S)Nutritious plant peptone containing high carbohydrate and vitamin contentCapable of supporting the growth of most fastidious bacteria and fungiCannot be used for fermentation studies due to its high carbohydrate contentProduces clear solutions at culture media concentration and neutral reactions after autoclavingProvides nitrogen, vitamins, and minerals in prepared culture mediaDehydrated Appearance: beige, homogeneous powderAnimal-free: No animal products.pH (2% solution at 25° C): 6.5 - 7.5From papaic hydrolysis of soybean meal.* Low NaCl content.* Total Nitrogen (TN) 8.7%* Amino Nitrogen (AN) 2.3%* AN:TN Ratio 0.26* Soluble in water
Catalog No. 50-254-349 Supplier Bioworld Supplier No. 306200611
Papaic Digest of Soybean Meal (Peptone S)Nutritious plant peptone containing high carbohydrate and vitamin contentCapable of supporting the growth of most fastidious bacteria and fungiCannot be used for fermentation studies due to its high carbohydrate contentProduces clear solutions at culture media concentration and neutral reactions after autoclavingProvides nitrogen, vitamins, and minerals in prepared culture mediaDehydrated Appearance: beige, homogeneous powderAnimal-free: No animal products.pH (2% solution at 25° C): 6.5 - 7.5From papaic hydrolysis of soybean meal.* Low NaCl content.* Total Nitrogen (TN) 8.7%* Amino Nitrogen (AN) 2.3%* AN:TN Ratio 0.26* Soluble in water