Learn More
Worthington Biochemical Corporation Papain, Lyophilized, 100mg
SDP
Catalog No. NC9912923
Encompass
Supplied as a lyophilized powder prepared from a two times crystalline suspension, Code: PAP. To insure full activity, the enzyme should be incubated in a solution containing 1.1mM EDTA, 0.067mM mercaptoethanol and 5.5mM cysteine-HCl for 30 minutes. It is recommended that the enzyme be 0.22 micron filtered after dissolution and prior to use. Store at 2-8°C. Papain is a sulfhydryl protease from Carica papaya latex. It has a molecular weight of 23 kDa and an optimum pH range of 6.0-7.0. The action of papain on leucine methyl ester produces an insoluble polyleucine peptide. Papain breaks down the intercellular matrix of cartilage. Papain is activated by cysteine, sulfide, and sulfite.
Catalog No. NC9912923 Supplier Worthington Biochemical Corporation Supplier No. LS003119
Supplied as a lyophilized powder prepared from a two times crystalline suspension, Code: PAP. To insure full activity, the enzyme should be incubated in a solution containing 1.1mM EDTA, 0.067mM mercaptoethanol and 5.5mM cysteine-HCl for 30 minutes. It is recommended that the enzyme be 0.22 micron filtered after dissolution and prior to use. Store at 2-8°C. Papain is a sulfhydryl protease from Carica papaya latex. It has a molecular weight of 23 kDa and an optimum pH range of 6.0-7.0. The action of papain on leucine methyl ester produces an insoluble polyleucine peptide. Papain breaks down the intercellular matrix of cartilage. Papain is activated by cysteine, sulfide, and sulfite.