Learn More
Worthington Biochemical Corporation PAPAIN 25MG
SDP
Catalog No. NC9110851
Encompass
Supplied as a two times crystalline suspension in 50mM sodium acetate, pH 4.5. To insure full activity, the enzyme should be incubated in a solution containing 1.1mM EDTA, 0.067mM mercaptoethanol and 5.5mM cysteine-HCl for 30 minutes.
Catalog No. NC9110851 Supplier Worthington Biochemical Corporation Supplier No. LS003124
Supplied as a two times crystalline suspension in 50mM sodium acetate, pH 4.5. To insure full activity, the enzyme should be incubated in a solution containing 1.1mM EDTA, 0.067mM mercaptoethanol and 5.5mM cysteine-HCl for 30 minutes.