Learn More
Cayman Chemical Papain Inhibitor (trifluoroacetate salt), Purity: ≥95%, 100 mg
SDP
Catalog No. 502990946
Encompass_Preferred
A peptide inhibitor of papain (Ki = 113 µM)
Catalog No. 50-299-0946 Supplier Cayman Chemical Supplier No. 31404100 MG
A peptide inhibitor of papain (Ki = 113 µM)