Learn More
Cayman Chemical Papain Inhibitor (trifluoroacetate salt), Purity: ≥95%, 50 mg
SDP
Catalog No. 502991742
Encompass_Preferred
A peptide inhibitor of papain (Ki = 113 µM)
Catalog No. 50-299-1742 Supplier Cayman Chemical Supplier No. 3140450 MG
A peptide inhibitor of papain (Ki = 113 µM)