Learn More
Worthington Biochemical Corporation Papain, Lyophilized, 25mg
SDP
Catalog No. NC0056304
Encompass
Supplied as a lyophilized powder prepared from a two times crystalline suspension, Code: PAP. To insure full activity, the enzyme should be incubated in a solution containing 1.1mM EDTA, 0.067mM mercaptoethanol and 5.5mM cysteine-HCl for 30 minutes. It is recommended that the enzyme be 0.22 micron filtered after dissolution and prior to use. Store at 2-8°C.
Catalog No. NC0056304 Supplier Worthington Biochemical Corporation Supplier No. LS003118
Supplied as a lyophilized powder prepared from a two times crystalline suspension, Code: PAP. To insure full activity, the enzyme should be incubated in a solution containing 1.1mM EDTA, 0.067mM mercaptoethanol and 5.5mM cysteine-HCl for 30 minutes. It is recommended that the enzyme be 0.22 micron filtered after dissolution and prior to use. Store at 2-8°C.