Learn More
Sigma Aldrich Fine Chemicals Biosciences Papain from papaya latex, 9001-73-4, MFCD00131791, 25 mg
Catalog No. NC1052280
Encompass
Papain is a thermostable enzyme and becomes inactive at 55°C. It has two domains and the catalytic region is present at the domain interface. The cysteine 25 in the active site performs the proteolysis by nucleophilic attack on the carbonyl carbon of a peptides. Papain has wide industrial applications in food and pharmaceutical industry.
Catalog No. NC1052280 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P530625MG
Papain is a thermostable enzyme and becomes inactive at 55°C. It has two domains and the catalytic region is present at the domain interface. The cysteine 25 in the active site performs the proteolysis by nucleophilic attack on the carbonyl carbon of a peptides. Papain has wide industrial applications in food and pharmaceutical industry.