Learn More
Bioworld PAPAIN SEPAROPOREAGAROSE) 4B
Supplier:  Bioworld 201810272
Encompass
Specifications: Matrix: Cross-linked 4% Agarose beads , Separopore 4B; Activity: 16-40 BAEE units per milligram of papain before immobilization ; Loading: 250mg papain/ml of settled gel; Particle size (range): 63-150 m; Supplied as: ...
Catalog No. 50-197-731